Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

C62506

Sigma-Aldrich

1-Chloro-3-pentanone

technical grade, 85%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COCH2CH2Cl
CAS Number:
Molecular Weight:
120.58
Beilstein/REAXYS Number:
1699401
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

assay

85%

form

liquid

refractive index

n20/D 1.435 (lit.)

bp

68 °C/20 mmHg (lit.)

density

1.042 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCC(=O)CCCl

InChI

1S/C5H9ClO/c1-2-5(7)3-4-6/h2-4H2,1H3

InChI key

APNSUHRNUVUCIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

123.8 °F - closed cup

flash_point_c

51 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service