C122009
L-Cystine
99%
Synonym(s):
(R,R)-3,3′-Dithiobis(2-aminopropionic acid)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein/REAXYS Number:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352209
assay
99%
optical activity
[α]20/D −217.8°, c = 1 in 1 M HCl
mp
>240 °C (dec.) (lit.)
SMILES string
N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O
InChI
1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1
InChI key
LEVWYRKDKASIDU-IMJSIDKUSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Philip J Hogg
Nature reviews. Cancer, 13(6), 425-431 (2013-05-11)
Protein action in nature is generally controlled by the amount of protein produced and by chemical modification of the protein, and both are often perturbed in cancer. The amino acid side chains and the peptide and disulphide bonds that bind
Serge-George Rosolen et al.
Advances in experimental medicine and biology, 775, 145-154 (2013-02-09)
In the 70s, the amino acid taurine was found essential for photoreceptor survival. Recently, we found that taurine depletion can also trigger retinal ganglion cell degeneration both in vitro and in vivo. Therefore, evaluation of taurine levels could be a
Martijn J Wilmer et al.
American journal of physiology. Renal physiology, 299(5), F905-F916 (2010-09-10)
Renal proximal tubules are highly sensitive to ischemic and toxic insults and are affected in diverse genetic disorders, of which nephropathic cystinosis is the most common. The disease is caused by mutations in the CTNS gene, encoding the lysosomal cystine
Anne C Conibear et al.
The Journal of biological chemistry, 288(15), 10830-10840 (2013-02-23)
θ-Defensins are ribosomally synthesized cyclic peptides found in the leukocytes of some primate species and have promising applications as antimicrobial agents and scaffolds for peptide drugs. The cyclic cystine ladder motif, comprising a cyclic peptide backbone and three parallel disulfide
Jan Lewerenz et al.
Antioxidants & redox signaling, 18(5), 522-555 (2012-06-07)
The antiporter system x(c)(-) imports the amino acid cystine, the oxidized form of cysteine, into cells with a 1:1 counter-transport of glutamate. It is composed of a light chain, xCT, and a heavy chain, 4F2 heavy chain (4F2hc), and, thus
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service