About This Item
Recommended Products
assay
95%
mp
67-69 °C (lit.)
SMILES string
BrCCC(C#N)(c1ccccc1)c2ccccc2
InChI
1S/C16H14BrN/c17-12-11-16(13-18,14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-10H,11-12H2
InChI key
IGYSFJHVFHNOEI-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service