B35105
3,3′-(4,4′-Biphenylene)bis(2,5-diphenyl-2H-tetrazolium chloride)
Synonym(s):
2,2′,5,5′-Tetraphenyl-3,3′-(p-diphenylene)ditetrazolium chloride, NTC, NeoT, Neotetrazolium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C38H28Cl2N8
CAS Number:
Molecular Weight:
667.59
Beilstein/REAXYS Number:
3887530
EC Number:
MDL number:
UNSPSC Code:
12352200
mp
230 °C (dec.) (lit.)
λmax
250 nm
SMILES string
[Cl-].[Cl-].c1ccc(cc1)-c2nn(-c3ccccc3)[n+](n2)-c4ccc(cc4)-c5ccc(cc5)-[n+]6nc(nn6-c7ccccc7)-c8ccccc8
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service