B29202
Benzyl phenyl sulfide
99%
Synonym(s):
Benzylthiobenzene
About This Item
assay
99%
bp
197 °C/27 mmHg (lit.)
mp
40-44 °C (lit.)
SMILES string
C(Sc1ccccc1)c2ccccc2
Looking for similar products? Visit Product Comparison Guide
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service