A93097
DL-Aspartic acid
98%
Synonym(s):
(±)-2-Aminosuccinic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HOOCCH2CH(NH2)COOH
CAS Number:
Molecular Weight:
133.10
Beilstein/REAXYS Number:
774618
EC Number:
MDL number:
UNSPSC Code:
12352103
Recommended Products
assay
98%
mp
>300 °C (lit.)
SMILES string
NC(CC(O)=O)C(O)=O
InChI
1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)
InChI key
CKLJMWTZIZZHCS-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
replaced by
Product No.
Description
Pricing
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chromatograms
application for HPLCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service