Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A83202

Sigma-Aldrich

D-Amygdalin

Synonym(s):

D-Mandelonitrile β-gentiobioside, D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, D-Mandelonitrile-β-gentiobioside, Amygdalin, Laetrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H27NO11
CAS Number:
Molecular Weight:
457.43
Beilstein/REAXYS Number:
66856
EC Number:
MDL number:
UNSPSC Code:
12352103

optical activity

[α]22/D −38°, c = 1.2 in H2O

mp

223-226 °C (lit.)

SMILES string

OC[C@H]1O[C@@H](OC[C@H]2O[C@@H](OC(C#N)c3ccccc3)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Biochem/physiol Actions

Cyanogenic glycoside that is a component of bitter almonds and apricot pits. There is no scientific evidence that amygdalin itself is an effective anti-cancer agent. Recent studies using β−glucoside linked to a tumor-associated monoclonal antibody to release cyanide at the tumor cell has shown significant cytotoxicity.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James P Tauber et al.
Insects, 11(11) (2020-11-15)
Amygdalin, a cyanogenic glycoside, is found in the nectar and pollen of almond trees, as well as in a variety of other crops, such as cherries, nectarines, apples and others. It is inevitable that western honeybees (Apis mellifera) consistently consume
Ping Deng et al.
Foods (Basel, Switzerland), 10(2) (2021-03-07)
To reveal the accumulation pattern of cyanogenic glycosides (amygdalin and prunasin) in bitter apricot kernels to further understand the metabolic mechanisms underlying differential accumulation during kernel development and ripening and explore the association between cyanogenic glycoside accumulation and the physical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service