Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A51602

Sigma-Aldrich

2-Amino-6,7-dimethyl-4-hydroxypteridine

97% dry basis

Synonym(s):

2-Amino-6,7-dimethyl-4-pteridinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N5O
CAS Number:
Molecular Weight:
191.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97% dry basis

mp

>300 °C (lit.)

SMILES string

Cc1nc2nc(N)nc(O)c2nc1C

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emilia Daghir-Wojtkowiak et al.
Journal of chromatography. A, 1403, 54-62 (2015-06-04)
The objective of this study was to model the retention of nucleosides and pterins in hydrophilic interaction liquid chromatography (HILIC) via QSRR-based approach. Two home-made (Amino-P-C18, Amino-P-C10) and one commercial (IAM.PC.DD2) HILIC stationary phases were considered. Logarithm of retention factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service