Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A39300

Sigma-Aldrich

4-Aminoantipyrine

98%

Synonym(s):

4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, Ampyrone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O
CAS Number:
Molecular Weight:
203.24
Beilstein/REAXYS Number:
181635
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

105-110 °C (lit.)

SMILES string

CN1N(c2ccccc2)C(=O)C(N)=C1C

InChI

1S/C11H13N3O/c1-8-10(12)11(15)14(13(8)2)9-6-4-3-5-7-9/h3-7H,12H2,1-2H3

InChI key

RLFWWDJHLFCNIJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used in the colorimetric determination of phenols[1] and the synthesis of transition metal[2] and lanthanide ligands.[3]

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transition Met. Chem. (London), 18, 279-279 (1993)
Analytical Letters, 26, 87-87 (1993)
Polyhedron, 12, 1227-1227 (1993)
Qihui Wang et al.
Journal of hazardous materials, 186(2-3), 1076-1081 (2010-12-21)
In this work, the immobilization of 4-aminoantipyrine onto bentonite was carried out and it was then used to investigate the adsorption behavior of Cr(III), Hg(II) and Pb(II) ions from aqueous solutions. The separation and preconcentration conditions of analytes were investigated
Mohammad Sayed Alam et al.
Bioorganic & medicinal chemistry, 20(13), 4103-4108 (2012-05-26)
4-Aminoantipyrine (4-amino-1,5-dimethyl-2-phenylpyrazole-3-one) and its analogues have been found to be compounds of interest for their anti-inflammatory, analgesic, antiviral, antipyretic, antirheumatic and antimicrobial activities. In the present study, Schiff base analogues of 4-aminoantipyrine were synthesized by the condensation reaction with substituted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service