Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

94424

Sigma-Aldrich

4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heptadecafluoroundecanoic acid potassium salt

≥97.0% (NT)

Synonym(s):

2H,2H,3H,3H-Perfluoroundecanoic acid potassium salt, Potassium 4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-heptadecafluoroundecanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7CH2CH2COOK
CAS Number:
Molecular Weight:
530.22
Beilstein/REAXYS Number:
7509593
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0% (NT)

mp

~210 °C (dec.)

SMILES string

[K+].[O-]C(=O)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Lact. - Repr. 1B - STOT RE 1

target_organs

Liver

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service