Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92036

Sigma-Aldrich

Triisobutylphosphine sulfide

≥97.0% (CHN)

Synonym(s):

TIBPS

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHCH2]3PS
CAS Number:
Molecular Weight:
234.38
Beilstein/REAXYS Number:
1762664
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥97.0% (CHN)

mp

57-60 °C

SMILES string

CC(C)CP(=S)(CC(C)C)CC(C)C

InChI

1S/C12H27PS/c1-10(2)7-13(14,8-11(3)4)9-12(5)6/h10-12H,7-9H2,1-6H3

InChI key

MXRLZCWBOJMFJG-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rafael J González-Álvarez et al.
Membranes, 10(10) (2020-10-09)
Popularity of hollow fiber-supported liquid membranes (HF-SLM) for liquid-phase microextraction (HF-LPME) has increased in the last decades. In particular, HF-SLM are applied for sample treatment in the determination and speciation of metals. Up to the date, optimization of preconcentration systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service