About This Item
assay
≥97.0%
refractive index
n20/D 1.446
density
0.806 g/mL at 20 °C (lit.)
SMILES string
CCCCCCCN(CCCCCCC)CCCCCCC
InChI
1S/C21H45N/c1-4-7-10-13-16-19-22(20-17-14-11-8-5-2)21-18-15-12-9-6-3/h4-21H2,1-3H3
InChI key
CLZGJKHEVKJLLS-UHFFFAOYSA-N
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service