Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91739

Sigma-Aldrich

1-(Trifluoromethanesulfonyl)imidazole

technical, ≥95.0% (RT)

Synonym(s):

1-(Trifluoromethylsulfonyl)imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3F3N2O2S
CAS Number:
Molecular Weight:
200.14
Beilstein/REAXYS Number:
912561
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

grade

technical

assay

≥95.0% (RT)

refractive index

n20/D 1.416

mp

19-21 °C

density

1.525 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)S(=O)(=O)n1ccnc1

InChI

1S/C4H3F3N2O2S/c5-4(6,7)12(10,11)9-2-1-8-3-9/h1-3H

InChI key

YGABUCCNCBMODG-UHFFFAOYSA-N

Other Notes

Electrophilic trifyl source. Review[1]; Preparation of vinyltriflates. Review[2]

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.J. Stang
Accounts of Chemical Research, 11, 107-107 (1978)
J.B. Hendrickson et al.
Organic preparations and procedures international, 9, 173-173 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service