Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87901

Sigma-Aldrich

(S)-3-(Fmoc-amino)-6-phenyl-5-hexenoic acid

≥97.0% (HPLC)

Synonym(s):

Fmoc-4-styryl-L-β-homoalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25NO4
CAS Number:
Molecular Weight:
427.49
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97.0% (HPLC)

storage temp.

2-8°C

SMILES string

OC(=O)C[C@H](C\C=C\c1ccccc1)NC(=O)OCC2c3ccccc3-c4ccccc24


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service