Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86898

Sigma-Aldrich

Tetrabutylammonium sulfamate

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(NH2SO3)
CAS Number:
Molecular Weight:
338.55
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

≥97.0%

SMILES string

NS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Other Notes

This lipophilic sulfamate can be useful for many purposes, e.g. the synthesis of N-acyl sulfamates (amide N-sulfonates) from a carboxylic acid; used in the preparation of monobactams

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.M. Cimarusti et al.
Tetrahedron, 39, 2577-2577 (1983)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service