Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

86878

Sigma-Aldrich

Tetrabutylammonium hydrogen sulfide

technical, ≥90% (NT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(SH)
CAS Number:
Molecular Weight:
275.54
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical

assay

≥90% (NT)

storage temp.

2-8°C

SMILES string

[SH-].CCCC[N+](CCCC)(CCCC)CCCC


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service