Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

862266

Sigma-Aldrich

D-Altrose

97%

Synonym(s):

Alt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Beilstein/REAXYS Number:
1724621
EC Number:
UNSPSC Code:
12352103
Pricing and availability is not currently available.

Quality Level

assay

97%

SMILES string

OC[C@H]1OC(O)[C@@H](O)[C@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,3-6,8-12H,2H2

InChI key

GZCGUPFRVQAUEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke L Lairson et al.
Nature chemical biology, 2(12), 724-728 (2006-10-24)
Despite their unparalleled catalytic prowess and environmental compatibility, enzymes have yet to see widespread application in synthetic chemistry. This lack of application and the resulting underuse of their enormous potential stems not only from a wariness about aqueous biological catalysis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service