Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

862002

Sigma-Aldrich

DL-Cycloserine

97%

Synonym(s):

4-Amino-3-isoxazolidinone, 4-Amino-3-isoxazolidone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6N2O2
CAS Number:
Molecular Weight:
102.09
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

97%

SMILES string

NC1CONC1=O

InChI

1S/C3H6N2O2/c4-2-1-7-5-3(2)6/h2H,1,4H2,(H,5,6)

InChI key

DYDCUQKUCUHJBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of alanine aminotransferase and other pyridoxal-phosphate-requiring transaminases; blocks sphingosine biosynthesis by inhibition of ketosphinganine synthetase.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service