Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

861391

Sigma-Aldrich

Trioxsalen

98%

Synonym(s):

2,5,9-Trimethylfuro[3,2-g]benzopyran-7-one, 4,5′,8-Trimethylpsoralen, TMP, Trisoralen

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O3
CAS Number:
Molecular Weight:
228.24
Beilstein/REAXYS Number:
221723
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

229-231 °C (lit.)

SMILES string

Cc1cc2cc3C(C)=CC(=O)Oc3c(C)c2o1

InChI

1S/C14H12O3/c1-7-4-12(15)17-14-9(3)13-10(6-11(7)14)5-8(2)16-13/h4-6H,1-3H3

InChI key

FMHHVULEAZTJMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Photochemical crosslinker of DNA that has been used as a probe for nucleic acid structure and function.
Photochemical crosslinker of DNA that has been used as a probe for nucleic acid structure and function. Trioxsalen has also been used to crosslink DNA onto mica surfaces.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service