Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

860859

Sigma-Aldrich

Citrinin

98%

Synonym(s):

4,6-Dihydro-8-hydroxy-3,4,5-trimethyl-6-oxo-3H-2-benzopyran-7-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14O5
CAS Number:
Molecular Weight:
250.25
Beilstein/REAXYS Number:
5282243
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

assay

98%

SMILES string

C[C@H]1OC=C2C(O)=C(C(O)=O)C(=O)C(C)=C2[C@@H]1C

InChI

1S/C13H14O5/c1-5-7(3)18-4-8-9(5)6(2)11(14)10(12(8)15)13(16)17/h4-5,7,15H,1-3H3,(H,16,17)/t5-,7-/m1/s1

InChI key

CQIUKKVOEOPUDV-IYSWYEEDSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuqiang Liu et al.
Toxicology in vitro : an international journal published in association with BIBRA, 26(6), 856-861 (2012-05-09)
A previous study has shown that CTN (Citrinin) inhibits mouse testosterone production. In this study, the mechanism by which testosterone production is inhibited by CTN in rat Leydig cells was investigated, and the morphological evidence of apoptosis, including nuclei fragmentation
Yishan Yang et al.
Fungal biology, 116(2), 225-233 (2012-02-01)
FlbA (fluffy low brlA expression), a regulator of the G protein signalling (RGS) pathway, has been implicated in the control of hyphal development, sporulation, mycotoxin/pigment production in many kinds of filamentous fungi and yeasts. In the current study, a FlbA-like
Mohammad Rafiqul Islam et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(10), 3537-3547 (2012-07-10)
The mycotoxin citrinin can cause mycotoxic nephropathy, cytotoxicity and genotoxicity. To investigate the immune modulatory effects, CTN was orally administered to female BALB/c mice at the dose of 1, 5, or 10 mg/kg body weight for 14 days, and several
Hassan Hajjaj et al.
Journal of food science, 77(3), M156-M159 (2012-03-06)
Amino acids were used as sole nitrogen sources to examine their effects on the production of water-soluble red pigments and citrinin by Monascus ruber ATCC 96218 cultivated on chemically defined media. In general, when glycine, tyrosine, arginine, serine, or histidine
H K Frank
Zeitschrift fur Ernahrungswissenschaft, 31(3), 164-177 (1992-09-01)
Citrinin, a nephrotoxic mycotoxin, has been of growing importance also for the "International Agency for Research on Cancer", ever since its presumable role in the occurrence of Balcan endemic nephropathy (BEN) was discussed at the congress on "Mycotoxins, Endemic Nepthropathy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service