Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

859958

Sigma-Aldrich

Cytochalasin D

from Zygosporium mansonii

Synonym(s):

Zygosporin A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H37NO6
CAS Number:
Molecular Weight:
507.62
Beilstein/REAXYS Number:
1632828
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

biological source

Zygosporium mansonii

Quality Level

SMILES string

[H][C@@]12[C@H](C)C(=C)[C@@H](O)[C@@H]3\C=C\C[C@H](C)C(=O)[C@](C)(O)\C=C\[C@@H](OC(C)=O)[C@@]13C(=O)N[C@H]2Cc4ccccc4

InChI

1S/C30H37NO6/c1-17-10-9-13-22-26(33)19(3)18(2)25-23(16-21-11-7-6-8-12-21)31-28(35)30(22,25)24(37-20(4)32)14-15-29(5,36)27(17)34/h6-9,11-15,17-18,22-26,33,36H,3,10,16H2,1-2,4-5H3,(H,31,35)/b13-9+,15-14+/t17-,18+,22-,23-,24+,25-,26+,29+,30+/m0/s1

InChI key

SDZRWUKZFQQKKV-JHADDHBZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent inhibitor of actin polymerization; disrupts actin microfilaments; activates the p53-dependent pathways; inhibits smooth muscle contraction; inhibits insulin-stimulated glucose transport.

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer S N Verhoekx et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 31(2), 328-334 (2012-09-19)
Matrix metalloproteinases (MMPs) are expressed in Dupuytren's contracture and play a role in matrix remodeling. We tested the role of tension on contractility and MMP expression in Dupuytren's nodule and cord cells. Cells were subjected to pre-determined loading patterns of
Marie Olliver et al.
The Journal of infectious diseases, 208(9), 1474-1481 (2013-08-08)
Streptococcus pneumoniae forms part of the normal nasopharyngeal flora but can also cause a broad spectrum of inflammatory diseases. Vitamin D has potent effects on human immunity, including induction of antimicrobial peptides and suppression of T-cell proliferation, but its ability
Katsuhiro Sasaki et al.
Nature communications, 6, 7484-7484 (2015-06-24)
Positive selection in the thymus provides low-affinity T-cell receptor (TCR) engagement to support the development of potentially useful self-major histocompatibility complex class I (MHC-I)-restricted T cells. Optimal positive selection of CD8(+) T cells requires cortical thymic epithelial cells that express
Pere Roca-Cusachs et al.
Biophysical journal, 94(12), 4984-4995 (2008-03-11)
Shape-dependent local differentials in cell proliferation are considered to be a major driving mechanism of structuring processes in vivo, such as embryogenesis, wound healing, and angiogenesis. However, the specific biophysical signaling by which changes in cell shape contribute to cell
Sahar Javaherian et al.
Integrative biology : quantitative biosciences from nano to macro, 7(3), 298-312 (2015-01-23)
During development and in adult tissues separation of phenotypically distinct cell populations is necessary to ensure proper organization and function of tissues and organs. Various phenomena, such as differential adhesion, differential mechanical tension and cell-cell repulsion, are proposed to cause

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service