Skip to Content
MilliporeSigma
All Photos(1)

Documents

859567

Sigma-Aldrich

p-Methoxybenzyl S-(4,6-dimethylpyrimidin-2-yl) thiocarbonate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16N2O3S
CAS Number:
Molecular Weight:
304.36
MDL number:
UNSPSC Code:
12352100

Quality Level

assay

98%

SMILES string

COc1ccc(COC(=O)Sc2nc(C)cc(C)n2)cc1

InChI

1S/C15H16N2O3S/c1-10-8-11(2)17-14(16-10)21-15(18)20-9-12-4-6-13(19-3)7-5-12/h4-8H,9H2,1-3H3

InChI key

DDIJXDDZBQMPDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service