858811
5-Bromo-2′-deoxyuridine
97%
Synonym(s):
5-BrdU, 5-Bromo-1-(2-deoxy-β-D-ribofuranosyl)uracil, 5-Bromouracil deoxyriboside, BUdR
About This Item
Recommended Products
assay
97%
optical activity
[α]22/D +23.0°, c = 1 in H2O
mp
191-194 °C (dec.) (lit.)
storage temp.
−20°C
SMILES string
OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(Br)C(=O)NC2=O
InChI
1S/C9H11BrN2O5/c10-4-2-12(9(16)11-8(4)15)7-1-5(14)6(3-13)17-7/h2,5-7,13-14H,1,3H2,(H,11,15,16)/t5-,6+,7+/m0/s1
InChI key
WOVKYSAHUYNSMH-RRKCRQDMSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
replaced by
signalword
Danger
hcodes
Hazard Classifications
Muta. 1B - Repr. 2
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Which document(s) contains shelf-life or expiration date information for a given product?
If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.
How do I get lot-specific information or a Certificate of Analysis?
The lot specific COA document can be found by entering the lot number above under the "Documents" section.
How do I find price and availability?
There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote. USA customers: 1-800-325-3010 or view local office numbers.
What is the stability of Product 858811, 5-Bromo-2´-deoxyuridine (BrdU)?
The comparable Sigma product, B5002, has a recommended retest specification of 2 years. We have observed by TLC that the material is unchanged after 3 years. Solutions at neutral pH should be stable if stored frozen for up to 6 months.
What can I use to detect BrdU, 5-Bromo-2´-deoxyuridine, in vivo and in vitro?
For the immunohistochemical detection of BrdU, we recommend the monoclonal anti-BrdU antibody, Product No. B2531.
What are the hazards associated with Product 858811, 5-Bromo-2´-deoxyuridine (BrdU)?
BrdU is a known carcinogen, and caution should be exercised when handling. Please refer to the MSDS.
How long do I treat cells with Product 858811, 5-Bromo-2´-deoxyuridine (BrdU) to incorporate it into the DNA?
Actively growing cells may be treated with 10 μM BrdU for 1 hour.
How does Product 858811, 5-Bromo-2´-deoxyuridine (BrdU), work as a thymidine analog?
BrdU replaces thymidine during the S-phase of cell division.
What is the Department of Transportation shipping information for this product?
Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
My question is not addressed here, how can I contact Technical Service for assistance?
Ask a Scientist here.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service