Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

858617

Sigma-Aldrich

N-(Iodoacetaminoethyl)-1-naphthylamine-5-sulfonic acid

Synonym(s):

1,5-I-AEDANS, 5-[2-(Iodoacetamido)ethylamino]naphthalene-1-sulfonic acid, N-Iodoacetyl-N′-(5-sulfo-1-naphthyl)ethylenediamine, Hudson-Weber-Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ICH2CONHCH2CH2NHC10H6SO3H
CAS Number:
Molecular Weight:
434.25
Beilstein/REAXYS Number:
3004454
EC Number:
MDL number:
UNSPSC Code:
12352103

SMILES string

OS(=O)(=O)c1cccc2c(NCCNC(=O)CI)cccc12

InChI

1S/C14H15IN2O4S/c15-9-14(18)17-8-7-16-12-5-1-4-11-10(12)3-2-6-13(11)22(19,20)21/h1-6,16H,7-9H2,(H,17,18)(H,19,20,21)

InChI key

ZMERMCRYYFRELX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for sulfhydryl determination

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service