Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

857254

Sigma-Aldrich

L-1-p-Tosylamino-2-phenylethyl chloromethyl ketone

≥99%

Synonym(s):

(S)-1-Chloro-3-tosylamido-4-phenyl-2-butanone, (S)-1-Chloro-4-phenyl-3-tosylamido-2-butanone, N-p-Tosyl-L-phenylalanine chloromethyl ketone, TPCK, Tosyl-L-phenylalanyl-chloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH(NHSO2C6H4CH3)COCH2Cl
CAS Number:
Molecular Weight:
351.85
Beilstein/REAXYS Number:
2895215
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

≥99%

mp

106-108 °C (lit.)

SMILES string

Cc1ccc(cc1)S(=O)(=O)N[C@@H](Cc2ccccc2)C(=O)CCl

Application

Protease inhibitor.

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service