Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

856231

Sigma-Aldrich

Aflatoxin G2

crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein/REAXYS Number:
1692017
MDL number:

form

crystalline

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)OCC5)C(=O)Oc4c23

InChI

1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WPCVRWVBBXIRMA-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

Warning

Possibly carcinogenic.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service