Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

855820

Sigma-Aldrich

Hexadecyltrimethylammonium bromide

95%

Synonym(s):

CTAB, Cetrimonium bromide, Cetyltrimethylammonium bromide, Palmityltrimethylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15N(Br)(CH3)3
CAS Number:
Molecular Weight:
364.45
Beilstein/REAXYS Number:
3598189
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

95%

mp

248-251 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCCCC[N+](C)(C)C

InChI

1S/C19H42N.BrH/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(2,3)4;/h5-19H2,1-4H3;1H/q+1;/p-1

InChI key

LZZYPRNAOMGNLH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Phase-transfer catalyst for polyamide,[1] polycarbonate,[2] and polythiocarbonate[2] formation.

replaced by

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2 Oral - STOT SE 3

target_organs

Gastrointestinal tract, Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

471.2 °F - closed cup

flash_point_c

244 °C - closed cup

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Macromol. Sci., Pure Appl. Chem., A31, 283-283 (1994)
Macromolecules, 27, 1087-1087 (1994)
Yin-Ming Li et al.
Environmental technology, 35(17-20), 2556-2568 (2014-08-26)
This study was carried out to develop a cost-effective and practicable sorbent for application in abrupt perfluorooctane sulphonate (PFOS) pollution accidents. The main merit of this work was that a waste material, namely construction and demolition (C&D) waste, was employed
Lobna Abdel Aziz Hussein et al.
Journal of AOAC International, 97(4), 1175-1182 (2014-08-26)
This work deals with spectrophotometric, spectrofluorimetric, and potentiometric analyses of cetyltrimethylammonium bromide (CTAB) cationic detergent. The spectrophotometric procedure depends on measuring the absorbance of its binary complex with eosin yellow in Britton-Robinson buffer (pH 4) at Lambda max 547 nm
Liming Zhang et al.
Journal of biomedical nanotechnology, 10(8), 1440-1449 (2014-07-16)
Although gold nanorods (GNRs) have been investigated extensively for optical hyperthermia therapies, the synthesis of rods is far from ideal. In this report, we optimized the synthesis of gold nanorods using hydroquinone as a reducing agent. Compared with the GNRs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service