Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

855804

Sigma-Aldrich

N-Acetyl-DL-tryptophan

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O3
CAS Number:
Molecular Weight:
246.26
Beilstein/REAXYS Number:
89478
EC Number:
MDL number:
UNSPSC Code:
12352209

assay

≥99%

mp

204-206 °C (dec.) (lit.)

SMILES string

CC(=O)NC(Cc1c[nH]c2ccccc12)C(O)=O

InChI

1S/C13H14N2O3/c1-8(16)15-12(13(17)18)6-9-7-14-11-5-3-2-4-10(9)11/h2-5,7,12,14H,6H2,1H3,(H,15,16)(H,17,18)

InChI key

DZTHIGRZJZPRDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Replaced by

Product No.
Description
Pricing

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiro Takayama et al.
Analytica chimica acta, 898, 73-84 (2015-11-04)
Chiral metabolites are found in a wide variety of living organisms and some of them are understood to be physiologically active compounds and biomarkers. However, the overall analysis of chiral metabolomics is quite difficult due to the high number of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service