Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

852554

Sigma-Aldrich

6-(Dimethylamino)purine

98%

Synonym(s):

N6,N6-Dimethyladenine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9N5
CAS Number:
Molecular Weight:
163.18
Beilstein/REAXYS Number:
7634
EC Number:
MDL number:
UNSPSC Code:
12352200

assay

98%

mp

259-262 °C (lit.)

SMILES string

CN(C)c1ncnc2[nH]cnc12

InChI

1S/C7H9N5/c1-12(2)7-5-6(9-3-8-5)10-4-11-7/h3-4H,1-2H3,(H,8,9,10,11)

InChI key

BVIAOQMSVZHOJM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masako Okamoto et al.
Bioorganic & medicinal chemistry, 19(1), 486-489 (2010-12-08)
Apoptosis signal-regulating kinase 1 (ASK1, also called MAP3K5) is a mitogen-activated protein kinase kinase kinase (MAP3K) that plays important roles in stress-induced cell death and inflammation, and is expected as a new therapeutic target for cancer, cardiovascular diseases, and neurodegenerative
Oleg Fedorov et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(51), 20523-20528 (2007-12-14)
Protein kinases play a pivotal role in cell signaling, and dysregulation of many kinases has been linked to disease development. A large number of kinase inhibitors are therefore currently under investigation in clinical trials, and so far seven inhibitors have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service