Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

852139

Sigma-Aldrich

Uridine 5′-triphosphate trisodium salt dihydrate

Synonym(s):

UTP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C9H12N2O15P3Na3
CAS Number:
Molecular Weight:
586.12
Beilstein/REAXYS Number:
3585234
EC Number:
MDL number:
UNSPSC Code:
12352200

impurities

≤5% ethyl alcohol

mp

>140 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])(=O)OP(O)([O-])=O)N2C=CC(=O)NC2=O

InChI

1S/C9H15N2O15P3.3Na.2H2O/c12-5-1-2-11(9(15)10-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H,10,12,15)(H2,16,17,18);;;;2*1H2/q;3*+1;;/p-3/t4-,6-,7-,8-;;;;;/m1...../s1

InChI key

KIYVKLDQLQZXKH-GTIFRNKBSA-K

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

P2Y receptor agonist.

Other Notes

Contains methyl alcohol

replaced by


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service