Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

851582

Sigma-Aldrich

Xylitol

98%

Synonym(s):

Xylite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]3CH2OH
CAS Number:
Molecular Weight:
152.15
Beilstein/REAXYS Number:
1720523
EC Number:
MDL number:
UNSPSC Code:
12352103

assay

98%

mp

94-97 °C (lit.)

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+

InChI key

HEBKCHPVOIAQTA-SCDXWVJYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service