Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83903

Sigma-Aldrich

Ricinoleic acid

technical, ~80% (GC)

Synonym(s):

(R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(OH)CH2CH=CH(CH2)7COOH
CAS Number:
Molecular Weight:
298.46
Beilstein/REAXYS Number:
1727813
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

technical

assay

~80% (GC)

density

0.940 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

InChI

1S/C18H34O3/c1-2-3-4-11-14-17(19)15-12-9-7-5-6-8-10-13-16-18(20)21/h9,12,17,19H,2-8,10-11,13-16H2,1H3,(H,20,21)/b12-9-/t17-/m1/s1

InChI key

WBHHMMIMDMUBKC-QJWNTBNXSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

435.2 °F - closed cup

flash_point_c

224 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ioannis Mavraganis et al.
Applied and environmental microbiology, 76(4), 1135-1142 (2009-12-22)
Claviceps purpurea, the fungal pathogen that causes the cereal disease ergot, produces glycerides that contain high levels of ricinoleic acid [(R)-12-hydroxyoctadec-cis-9-enoic acid] in its sclerotia. Recently, a fatty acid hydroxylase (C. purpurea FAH [CpFAH]) involved in the biosynthesis of ricinoleic
Ming Li Wang et al.
Journal of agricultural and food chemistry, 59(17), 9250-9256 (2011-08-16)
Castor has tremendous potential as a feedstock for biodiesel production. The oil content and fatty acid composition in castor seed are important factors determining the price for production and affecting the key fuel properties of biodiesel. There are 1033 available
Bruno Rodrigues Sampieri et al.
Microscopy research and technique, 75(5), 683-690 (2011-12-02)
Rhipicephalus sanguineus is a widely distributed tick species that has adapted to the urban environment, and the dog is its main host. This species is also known as a vector and reservoir of diseases caused by bacteria, protozoa, and viruses.
André Arnosti et al.
Experimental parasitology, 127(2), 575-580 (2010-11-13)
This study examines the effects of ricinoleic acid esters from Ricinus communis castor oil on the vitellogenesis of Rhipicephalus sanguineus ticks attached to hosts that were fed with commercial rabbit food containing these esters. The oocytes of ticks from the
Petra Kocbek et al.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences, 50(1), 130-138 (2013-04-23)
Superparamagnetic iron oxide nanoparticles (SPIONs) show a great promise for a wide specter of bioapplications, due to their characteristic magnetic properties exhibited only in the presence of magnetic field. Their advantages in the fields of magnetic drug targeting and imaging

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service