Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

799491

Sigma-Aldrich

Conjugated microporous polyaryleneethynylene copolymer

Synonym(s):

CMP-1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C21H9)n
UNSPSC Code:
26111700
NACRES:
NA.23

form

crystals

Application

CMP-1 is a conjugated microporous material with applications in molecular separation, heterogeneous catalysis and gas storage.[1][2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia-Xing Jiang et al.
Journal of the American Chemical Society, 130(24), 7710-7720 (2008-05-27)
A series of rigid microporous poly(aryleneethynylene) (PAE) networks was synthesized by Sonogashira-Hagihara coupling chemistry. PAEs with apparent Brunauer-Emmet-Teller surface areas of more than 1000 m(2)/g were produced. The materials were found to have very good chemical and thermal stability and
R. Dawson
Macromolecules, 43, 8524-8524 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service