Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

79705

Sigma-Aldrich

3,5-Dimethylisoxazole

Wacker Chemie AG, ≥98.0% (GC)

Synonym(s):

Dimexazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO
CAS Number:
Molecular Weight:
97.12
Beilstein/REAXYS Number:
3935337
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.0% (GC)

manufacturer/tradename

Wacker Chemie AG

refractive index

n20/D 1.442 (lit.)

bp

142-144 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(C)on1

InChI

1S/C5H7NO/c1-4-3-5(2)7-6-4/h3H,1-2H3

InChI key

FICAQKBMCKEFDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service