Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

779970

Sigma-Aldrich

4-Chloro-3-fluoroaniline

≥97.0% (GC)

Synonym(s):

4-Chloro-3-fluorophenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5ClFN
CAS Number:
Molecular Weight:
145.56
Beilstein/REAXYS Number:
2716278
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (GC)
97.0-103.0% (NT)

form

crystals

functional group

chloro
fluoro

SMILES string

Nc1ccc(Cl)c(F)c1

InChI

1S/C6H5ClFN/c7-5-2-1-4(9)3-6(5)8/h1-3H,9H2

InChI key

ACMJJQYSPUPMPN-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

140.0 - 143.6 °F

flash_point_c

60 - 62 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service