Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

775711

Sigma-Aldrich

Manganese(IV) oxide

nanorod, diam. × L 5-30 nm × 80-100 nm, 99% trace metals basis (excludes potassium content)

Synonym(s):

Manganese dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MnO2
CAS Number:
Molecular Weight:
86.94
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

assay

99% trace metals basis (excludes potassium content)

form

nanorod

diam. × L

5-30 nm × 80-100 nm

mp

535 °C (dec.) (lit.)

SMILES string

O=[Mn]=O

InChI

1S/Mn.2O

Inchi Key

NUJOXMJBOLGQSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Manganese dioxide nanorods have been studied as supercapacitor, catalysts, and lithium ion batteries.

Pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Inhalation

target_organs

Brain

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han, J.; et al.
The Journal of Physical Chemistry C, 116, 15900-15907 (2012)
Sugantha, M.; et al.
International Journal of Hydrogen Energy, 28, 597-600 (2003)
Grigorieva, A.V.; et al.
Solid State Sciences, 14, 988-995 (2012)
Heng Lai et al.
ACS applied materials & interfaces, 4(5), 2325-2328 (2012-05-02)
MnO(2) nanoflakes coated on carbon nanohorns (CNHs) has been synthesized via a facile solution method and evaluated as anode for lithium-ion batteries. By using CNHs as buffer carrier, MnO(2)/CNH composite displays an excellent capacity of 565 mA h/g measured at
Wen-Hui Kuan et al.
Journal of hazardous materials, 239-240, 152-159 (2012-09-25)
This study examined the reaction of methylene blue (MB) with tunneled manganese oxide pyrolusite regarding pH and reaction time. MB was cleaved through N-demethylation, in which reaction azure B (AB), azure A (AA), azure C (AC), and thionin (TH) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service