Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75732

Sigma-Aldrich

3,5-Dimethoxy-α,α-dimethylbenzyl carbazate

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N2O4
CAS Number:
Molecular Weight:
254.28
Beilstein/REAXYS Number:
2465007
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

≥99.0%

mp

105-110 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

COc1cc(OC)cc(c1)C(C)(C)OC(=O)NN

InChI

1S/C12H18N2O4/c1-12(2,18-11(15)14-13)8-5-9(16-3)7-10(6-8)17-4/h5-7H,13H2,1-4H3,(H,14,15)

InChI key

RDYVDEBOFNOSEU-UHFFFAOYSA-N

Other Notes

The azide is prepared to introduce the Ddz protection, an N-protecting group labile toward weak acids and irradiation.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[The , -dimethyl-3,5-dimethoxybenzyloxycarbonyl (Ddz) residue, an N-protecting group labile toward weak acids and irradiation].
C Birr et al.
Justus Liebigs Annalen der Chemie, 763, 162-172 (1972-09-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service