Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

75525

Sigma-Aldrich

Diethyl phenyl orthoformate

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH(OC2H5)2OC6H5
CAS Number:
Molecular Weight:
196.24
Beilstein/REAXYS Number:
1872634
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥97.0%

form

liquid

refractive index

n20/D 1.480 (lit.)
n20/D 1.480

bp

103-104 °C/10 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)Oc1ccccc1

InChI

1S/C11H16O3/c1-3-12-11(13-4-2)14-10-8-6-5-7-9-10/h5-9,11H,3-4H2,1-2H3

InChI key

QTURWMMVIIBRRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the diethoxymethylation of Grignard reagents[1]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H. Stetter et al.
Chemische Berichte, 103, 643-643 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service