Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

740969

Sigma-Aldrich

Barium tetrafluorocobaltate

99.9% trace metals basis

Synonym(s):

Barium cobalt fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BaCoF4
CAS Number:
Molecular Weight:
272.25
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.9% trace metals basis
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

assay

99.9% trace metals basis

form

powder

impurities

≤2000.0 ppm Trace Metal Analysis

mp

>500 °C

SMILES string

[Ba++].F[Co--](F)(F)F

InChI

1S/Ba.Co.4FH/h;;4*1H/q2*+2;;;;/p-4

InChI key

HSUXUTLCZMELFQ-UHFFFAOYSA-J

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sun Woo Kim et al.
Journal of the American Chemical Society, 132(50), 17684-17685 (2010-12-01)
A low temperature, high yield hydrothermal route has been discovered for the phase-pure synthesis of the multiferroic BaMF(4) (M = Mg, Mn, Co, Ni, and Zn) family. The synthesis involves the use of CF(3)COOH instead of HF in an aqueous
J F Scott
Science (New York, N.Y.), 315(5814), 954-959 (2007-02-17)
Long viewed as a topic in classical physics, ferroelectricity can be described by a quantum mechanical ab initio theory. Thin-film nanoscale device structures integrated onto Si chips have made inroads into the semiconductor industry. Recent prototype applications include ultrafast switching
DiDomenico, Jr. M.; Eibshutz, M.; Guggenheim, H. J.; Camlibel, I.
Solid State Communications, 7, 1119-1119 (1969)
Ederer, C. and Spaldin, N. A.
Physical Review. B, Condensed Matter and Materials Physics, 74, 024102-024102 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service