Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

739502

Sigma-Aldrich

Tin(IV) tetrakis(trifluoromethanesulfonimide)

DMSO complex

Synonym(s):

1,1,1-Trifluoro-N-[(trifluoromethyl)sulfonyl]-N-[tris[bis[(trifluoromethyl)sulfonyl]amino]stannyl]-methanesulfonamide, Sn(IV) triflimate complex, Sn(NTf2)4-6·DMSO, Sn(NTf2)4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn[N[(SO2CF3)2]]4
CAS Number:
Molecular Weight:
1239.29
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: tin
reagent type: catalyst

SMILES string

CS(C)=O.CS(C)=O.CS(C)=O.CS(C)=O.CS(C)=O.CS(C)=O.FC(F)(F)S(=O)(=O)N([Sn](N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)(N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)N(S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F)S(=O)(=O)C(F)(F)F

InChI

1S/4C2F6NO4S2.6C2H6OS.Sn/c4*3-1(4,5)14(10,11)9-15(12,13)2(6,7)8;6*1-4(2)3;/h;;;;6*1-2H3;/q4*-1;;;;;;;+4

InChI key

SMVVMSYLGHHNFZ-UHFFFAOYSA-N

General description

May contain ca. 3-5% Free DMSO

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service