Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

738859

Sigma-Aldrich

Methyl 2-azidoacetate

97%

Synonym(s):

Methyl azidoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H5N3O2
CAS Number:
Molecular Weight:
115.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.440

density

1.182 g/mL at 25 °C

SMILES string

COC(=O)CN=[N+]=[N-]

InChI

1S/C3H5N3O2/c1-8-3(7)2-5-6-4/h2H2,1H3

InChI key

RXYCUKSOYJWGPE-UHFFFAOYSA-N

Application

  • Methyl 2-azidoacetate is widely used as a precursor to build triazole derivatives via alkyne-azide click chemistry.[1] Some of the examples include the synthesis of coumarin-triazole derivatives as potential antiplasmodial agents[2] and macrocyclic triazole containing largazole analogs as histone deacetylases-1 (HDAC1) inhibitors.[3]
  • It can be used to synthesize various pyrrole derivatives by Knoevenagel condensation[4] as in the synthesis of near-infrared boron dipyrromethene (BODIPY) donors for organic tandem solar cells.[5]
  • It can also be used in Hemetsberger-Knittel synthesis of substituted 5-, 6-, and 7-azaindoles.[6]
  • Synthesis of highly fluorescent pyreno[2,1-b]pyrroles using methyl 2-azidoacetate as one of the reagents.[7]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

120.0 °F

flash_point_c

48.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological evaluation of new largazole analogues: alteration of macrocyclic scaffold with click chemistry.
Li X, et al.
ACS Medicinal Chemistry Letters, 4(1), 132-136 (2012)
Highly fluorescent pyreno [2, 1-b] pyrroles: First syntheses, crystal structure, and intriguing photophysical properties.
Selvi S, et al.
The Journal of Organic Chemistry, 69(20), 6674-6678 (2004)
In vitro antiplasmodial efficacy of synthetic coumarin-triazole analogs.
Yadav N, et al.
European Journal of Medicinal Chemistry (2018)
Small Molecule Near-Infrared Boron Dipyrromethene Donors for Organic Tandem Solar Cells.
Li T Y, et al.
Journal of the American Chemical Society, 139(39), 13636-13639 (2017)
Transition metal-catalyzed synthesis of pyrroles from dienyl azides.
Dong H, et al.
Organic Letters, 9(25), 5191-5194 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service