Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

737399

Sigma-Aldrich

4-(Hydroxymethyl)-1-methylpiperidine

97%

Synonym(s):

(1-Methyl-4-piperidinyl)methanol, 4-(Hydroxymethyl)-N-methylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO
CAS Number:
Molecular Weight:
129.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.477

density

0.981 g/mL at 25 °C

SMILES string

CN1CCC(CO)CC1

InChI

1S/C7H15NO/c1-8-4-2-7(6-9)3-5-8/h7,9H,2-6H2,1H3

InChI key

KJZLJGZZDNGGCA-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F

flash_point_c

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service