Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

733474

Sigma-Aldrich

Dibutylboron triflate solution

1 M in toluene

Synonym(s):

Dibutylboryl trifluoromethanesulfonate solution, Dibutylboron triflate solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3B[(CH2)3CH3]2
CAS Number:
Molecular Weight:
274.11
Beilstein/REAXYS Number:
1968660
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

concentration

1 M in toluene

density

0.914 g/mL at 25 °C

SMILES string

CCCCB(CCCC)OS(=O)(=O)C(F)(F)F

InChI

1S/C9H18BF3O3S/c1-3-5-7-10(8-6-4-2)16-17(14,15)9(11,12)13/h3-8H2,1-2H3

InChI key

FAVAVMFXAKZTMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

3.2 °F - closed cup

flash_point_c

-16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service