Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

732605

Sigma-Aldrich

5-Amino-4-bromo-3-methylpyrazole hydrobromide

Synonym(s):

3-Amino-4-bromo-5-methyl-1H-pyrazole hydrobromide, 3-Amino-4-bromo-5-methylpyrazole hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6BrN3 · HBr
CAS Number:
Molecular Weight:
256.93
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

166-180 °C

functional group

bromo

SMILES string

Br.Cc1n[nH]c(N)c1Br

InChI

1S/C4H6BrN3.BrH/c1-2-3(5)4(6)8-7-2;/h1H3,(H3,6,7,8);1H

InChI key

UGCOULAJNKWOPX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service