Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

731781

Sigma-Aldrich

N-(Triphenylmethyl)methacrylamide

97%

Synonym(s):

2-Methyl-N-(triphenylmethyl)-2-propenamide, TrMAM

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H21NO
CAS Number:
Molecular Weight:
327.42
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

mp

130-135 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)NC(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C23H21NO/c1-18(2)22(25)24-23(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-17H,1H2,2H3,(H,24,25)

InChI key

UTYRFBWBJRYRSO-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sinuo Tan et al.
Journal of chromatography. A, 1625, 461298-461298 (2020-07-28)
This study has examined the batch binding behaviour of different thermo-responsive co-polymer grafted chromatographic materials under different temperature and protein loading conditions. The effect of molecular composition of poly(N-isopropylacrylamide) (PNIPAAm)-based co-polymers on the phase transition properties has been documented. Sixteen

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service