Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

730866

Sigma-Aldrich

Monodispersed silicon nanowires

undoped, diam. × L 150 nm ± 30 nm × 20 μm ± 2 μm, 1 x 106 wires/mL

Synonym(s):

NanoFET

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352103

form

dispersion

composition

, >99% (Silicon-basis)

diam. × L

150 nm ± 30 nm × 20 μm ± 2 μm

density

0.773 g/mL at 25 °C

SMILES string

[Si]

Looking for similar products? Visit Product Comparison Guide

General description

This material can be used in a variety of electronic and optical devices including transistors, photovoltaics, sensors and piezoelectric generators. They can be assembled onto different substrates, such as flexible or transparent ones amongst others.
Typically 1 μg/mL in Isopropyl alcohol

Legal Information

Product of Nanosys, Inc.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

53.6 °F

flash_point_c

12 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C McAlpine et al.
Nature materials, 6(5), 379-384 (2007-04-24)
The development of a robust method for integrating high-performance semiconductors on flexible plastics could enable exciting avenues in fundamental research and novel applications. One area of vital relevance is chemical and biological sensing, which if implemented on biocompatible substrates, could
Erik M Freer et al.
Nature nanotechnology, 5(7), 525-530 (2010-06-08)
Single-crystal nanowire transistors and other nanowire-based devices could have applications in large-area and flexible electronics if conventional top-down fabrication techniques can be integrated with high-precision bottom-up nanowire assembly. Here, we extend dielectrophoretic nanowire assembly to achieve a 98.5% yield of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service