Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72885

Sigma-Aldrich

4′-Nitrobenzo-18-crown-6

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23NO8
CAS Number:
Molecular Weight:
357.36
Beilstein/REAXYS Number:
1604291
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
Pricing and availability is not currently available.

assay

≥98.0%

mp

82-86 °C

SMILES string

[O-][N+](=O)c1ccc2OCCOCCOCCOCCOCCOc2c1

Other Notes

Reduction to the amine and used for preparing substituted benzo-18-crown-6[1][2]; Preparation of ring-cleaved derivatives[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G. Israel
J. Prakt. Chem., 324, 145-145 (1982)
L. Toke et al.
Liebigs Ann. Chem., 761-761 (1988)
S. Shinkai et al.
Journal of the American Chemical Society, 106, 1801-1801 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service