Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

727210

Sigma-Aldrich

Ethyl (R)-(−)-2-hydroxy-4-phenylbutyrate

ChiPros®, produced by BASF, 98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CH2CH(OH)CO2C2H5
CAS Number:
Molecular Weight:
208.25
Beilstein/REAXYS Number:
3590943
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

≥97.5% (GC)
98%

form

liquid

optical purity

enantiomeric excess: ≥97.5%

refractive index

n20/D 1.504 (lit.)

bp

212 °C (lit.)

density

1.075 g/mL at 20 °C (lit.)

SMILES string

CCOC(=O)[C@H](O)CCc1ccccc1

InChI

1S/C12H16O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7,11,13H,2,8-9H2,1H3/t11-/m1/s1

InChI key

ZJYKSSGYDPNKQS-LLVKDONJSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

302.0 °F - closed cup

flash_point_c

150 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chiral alcohols form a versatile class of chiral synthons, since they can be incorporated into the API structures directly as esters or ethers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service