Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726702

Sigma-Aldrich

(R)-4-Chloro-α-methylbenzylamine

ChiPros®, produced by BASF, ≥99%

Synonym(s):

(R)-1-(4-Chlorophenyl)ethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10ClN
CAS Number:
Molecular Weight:
155.62
Beilstein/REAXYS Number:
2412317
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

assay

≥99%
≥99.0% (GC)

form

liquid

optical purity

enantiomeric excess: ≥90.0%

refractive index

n20/D 1.543 (lit.)

density

1.110 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)c1ccc(Cl)cc1

InChI

1S/C8H10ClN/c1-6(10)7-2-4-8(9)5-3-7/h2-6H,10H2,1H3/t6-/m1/s1

InChI key

PINPOEWMCLFRRB-ZCFIWIBFSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

239.0 °F

flash_point_c

115 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service