Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

726680

Sigma-Aldrich

(R)-(−)-1-Cyclohexylethylamine

ChiPros®, produced by BASF, 99%

Synonym(s):

(R)-(−)-α-Methylcyclohexanemethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH(CH3)NH2
CAS Number:
Molecular Weight:
127.23
Beilstein/REAXYS Number:
2935070
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

produced by BASF

assay

≥98.5% (GC)
99%

form

liquid

optical activity

[α]20/D -3.8±0.3°, neat

optical purity

enantiomeric excess: ≥98.5%

bp

177-178 °C (lit.)

density

0.866 g/mL at 20 °C (lit.)

SMILES string

C[C@@H](N)C1CCCCC1

InChI

1S/C8H17N/c1-7(9)8-5-3-2-4-6-8/h7-8H,2-6,9H2,1H3/t7-/m1/s1

InChI key

XBWOPGDJMAJJDG-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

125.6 °F - closed cup

flash_point_c

52 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service